Learn More
Medchemexpress LLC V-9302 | 1855871-76-9 | 99.5% | 538.68 | 100 MG
SDP
Catalog No. 5000435610
Encompass_Preferred
V-9302 is a competitive antagonist of transmembrane glutamine flux. It selectively and potently targets the amino acid transporter ASCT2 (SLC1A5) but not ASCT1, inhibiting ASCT2-mediated glutamine uptake in HEK-293 cells.
  • Selectively targets the amino acid transporter ASCT2 (SLC1A5) over ASCT1.
  • Inhibits glutamine uptake (IC50=9.6 μM) in HEK-293 cells.
  • Pharmacological blockade of ASCT2 leads to attenuated cancer cell growth and proliferation, increases cell death, and increases oxidative stress.
  • Prevents tumor growth in HCT-116 and HT29 xenograft models (75 mg/kg; i.p.; daily for 21 days).
  • When combined with CB-839, it elicits strong growth inhibition in SNU398 and MHCC97H xenograft models.
  • High purity: 99.46%.
  • Intended for research use only, not for sale to patients.
Catalog No. 50-004-35610 Supplier Medchemexpress LLC Supplier No. HY112683100MG
V-9302 is a competitive antagonist of transmembrane glutamine flux. It selectively and potently targets the amino acid transporter ASCT2 (SLC1A5) but not ASCT1, inhibiting ASCT2-mediated glutamine uptake in HEK-293 cells.
  • Selectively targets the amino acid transporter ASCT2 (SLC1A5) over ASCT1.
  • Inhibits glutamine uptake (IC50=9.6 μM) in HEK-293 cells.
  • Pharmacological blockade of ASCT2 leads to attenuated cancer cell growth and proliferation, increases cell death, and increases oxidative stress.
  • Prevents tumor growth in HCT-116 and HT29 xenograft models (75 mg/kg; i.p.; daily for 21 days).
  • When combined with CB-839, it elicits strong growth inhibition in SNU398 and MHCC97H xenograft models.
  • High purity: 99.46%.
  • Intended for research use only, not for sale to patients.