Learn More
Selleck Chemical LLC V-9302 5mg 1855871-76-9
SDP
Supplier:  Selleck Chemical LLC S88185MG
Encompass_Preferred
V-9302 is a competitive small molecule antagonist of transmembrane glutamine flux, that selectively and potently targets the amino acid transporter ASCT2 (SLC1A5) with an IC50 value of 9.6 ?M for inhibition of glutamine uptake in HEK-293 cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8351