Learn More
Abcam V-9302, 1MG
Catalog No. 502680461
Encompass_Preferred
MW 538.7 Da, Purity >=98%. A competitive antagonist of transmembrane glutamine flux, that selectively targets the amino acid transporter ASCT2 (SLC1A5). V-9302 inhibits ASCT2-mediated glutamine uptake (IC₅₀ = 9.6 μM) in HEK-293 cells.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-0461 Supplier Abcam Supplier No. AB2870911MG
MW 538.7 Da, Purity >=98%. A competitive antagonist of transmembrane glutamine flux, that selectively targets the amino acid transporter ASCT2 (SLC1A5). V-9302 inhibits ASCT2-mediated glutamine uptake (IC₅₀ = 9.6 μM) in HEK-293 cells.
The product is subject to the following: Abcam Restricted Use Statement