Learn More
Selleck Chemical LLC V-9302 S8818-100MG
SDP
Catalog No. 5000025189
Encompass_Preferred
V-9302 is a competitive small molecule antagonist of transmembrane glutamine flux that selectively and potently targets the amino acid transporter ASCT2 (SLC1A5) with an IC50 value of 9 6 M for inhibition of glutamine uptake in HEK-293 cells V-9302 blocks Sodium-neutral AA transporter 2 (SNAT2 SLC38A2) and the large neutral AA transporter 1 (LAT1 SLC7A5) as observed in 143B osteosarcoma cells HCC1806 breast cancer cells and Xenopus laevis oocytes Dilution of PBS may cause a clear aqueous solution to turn into a uniform suspension
Catalog No. 50-000-25189 Supplier Selleck Chemical LLC Supplier No. S8818100MG
V-9302 is a competitive small molecule antagonist of transmembrane glutamine flux that selectively and potently targets the amino acid transporter ASCT2 (SLC1A5) with an IC50 value of 9 6 M for inhibition of glutamine uptake in HEK-293 cells V-9302 blocks Sodium-neutral AA transporter 2 (SNAT2 SLC38A2) and the large neutral AA transporter 1 (LAT1 SLC7A5) as observed in 143B osteosarcoma cells HCC1806 breast cancer cells and Xenopus laevis oocytes Dilution of PBS may cause a clear aqueous solution to turn into a uniform suspension