Learn More
Medchemexpress LLC HY-112683A 5mg Medchemexpress, V-9302 (hydrochloride) CAS:0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY112683A5MG
Encompass_Preferred
Medchemexpress, HY-112683A 5mg V-9302 (hydrochloride) CAS:0 V-9302 hydrochloride is a competitive antagonist of transmembrane glutamine flux. V-9302 hydrochloride selectively and potently targets the amino acid transporter ASCT2 (SLC1A5). V-9302 hydrochloride inhibits ASCT2-mediated glutamine uptake (IC50=9.6 µM) in HEK-293 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9735