Learn More
Grainger Dayton Wedge V-Belt, 3V Section, 118 in. Length, 1 ea.
Catalog No. 502773504
Encompass_Preferred
This 3V wedge V-belt is designed for efficient power transmission in single or multiple V-belt drive applications. Featuring a smaller top width and a deeper profile compared to classical belts, this wedge design enables higher power capacity. The belt is constructed from an abrasion-resistant rubber compound that provides durability against heat, oils, coolants, and ozone, while the polyester reinforcement cord ensures excellent stretch-resistance.
  • Wedge V-belt profile offers superior power transfer capabilities
  • Durable rubber compound construction resists high heat, oils, and ozone
  • Polyester cord reinforcement provides high strength and stretch resistance
  • Operating temperature range of -22°F to +194°F
  • Standard wedge design with 3/8 in. top width and 5/16 in. thickness
Catalog No. 50-277-3504 Supplier Grainger Supplier No. 13V731
This 3V wedge V-belt is designed for efficient power transmission in single or multiple V-belt drive applications. Featuring a smaller top width and a deeper profile compared to classical belts, this wedge design enables higher power capacity. The belt is constructed from an abrasion-resistant rubber compound that provides durability against heat, oils, coolants, and ozone, while the polyester reinforcement cord ensures excellent stretch-resistance.
  • Wedge V-belt profile offers superior power transfer capabilities
  • Durable rubber compound construction resists high heat, oils, and ozone
  • Polyester cord reinforcement provides high strength and stretch resistance
  • Operating temperature range of -22°F to +194°F
  • Standard wedge design with 3/8 in. top width and 5/16 in. thickness