Learn More
Grainger Dayton Wedge V-Belt, 3V Section, 30 in. Outside Length, 3/8 in. Top Width, 1 ea.
Catalog No. 502772267
Encompass_Preferred
This 3V wedge V-belt is designed for efficient power transmission, featuring a compact profile with a 3/8 in. top width and 5/16 in. thickness. Its wedge design enables higher power transfer compared to classical V-belts of an equivalent size, making it a reliable choice for general purpose applications. The belt is constructed from a robust rubber compound reinforced with polyester cords for durability and heat resistance.
  • Wedge V-belt design for optimized power transmission
  • Reinforced with polyester cord for heat and stretch resistance
  • Rubber compound material resists heat, oil, and ozone exposure
  • Operates in temperatures from -22°F to +194°F
  • 30 in. nominal outside length
  • Wrapped construction with a 3V section profile
Catalog No. 50-277-2267 Supplier Grainger Supplier No. 13V707
This 3V wedge V-belt is designed for efficient power transmission, featuring a compact profile with a 3/8 in. top width and 5/16 in. thickness. Its wedge design enables higher power transfer compared to classical V-belts of an equivalent size, making it a reliable choice for general purpose applications. The belt is constructed from a robust rubber compound reinforced with polyester cords for durability and heat resistance.
  • Wedge V-belt design for optimized power transmission
  • Reinforced with polyester cord for heat and stretch resistance
  • Rubber compound material resists heat, oil, and ozone exposure
  • Operates in temperatures from -22°F to +194°F
  • 30 in. nominal outside length
  • Wrapped construction with a 3V section profile