Learn More
Grainger Dayton Wedge V-Belt, 5V Section, 100 in. Length, 1 ea.
Catalog No. 502770761
Encompass_Preferred
This 5V wedge V-belt is designed to provide high power transmission capabilities for single or multiple V-belt drives. The belt features a deep profile and a specialized wedge design that allows it to transfer more power than classical V-belt alternatives of the same size. Constructed from an abrasion-resistant rubber compound, it delivers durability against flex fatigue and remains resistant to heat, oil, coolant, and ozone exposure. Reinforcing polyester cords ensure structural integrity and stability in general-purpose applications.
  • 5V wedge design for high power capacity
  • Operates in temperatures from -22°F to +194°F
  • Rubber compound resists heat, oils, coolants, and ozone
  • Strong polyester reinforcing cord for stretch resistance
  • 100 in. nominal outside length
  • 5/8 in. top width and 17/32 in. thickness
Catalog No. 50-277-0761 Supplier Grainger Supplier No. 13V747
This 5V wedge V-belt is designed to provide high power transmission capabilities for single or multiple V-belt drives. The belt features a deep profile and a specialized wedge design that allows it to transfer more power than classical V-belt alternatives of the same size. Constructed from an abrasion-resistant rubber compound, it delivers durability against flex fatigue and remains resistant to heat, oil, coolant, and ozone exposure. Reinforcing polyester cords ensure structural integrity and stability in general-purpose applications.
  • 5V wedge design for high power capacity
  • Operates in temperatures from -22°F to +194°F
  • Rubber compound resists heat, oils, coolants, and ozone
  • Strong polyester reinforcing cord for stretch resistance
  • 100 in. nominal outside length
  • 5/8 in. top width and 17/32 in. thickness