Learn More
Grainger Dayton Classical V-Belt, Wedge, 138 in. Length, 1 Rib
Catalog No. 502771472
Encompass_Preferred
This B-section classical V-belt features a wrapped construction designed for reliable power transmission in single or multiple V-belt drive applications. Engineered for performance in general industrial settings, the belt is built with polyester reinforcing cords and a rubber compound body to ensure durability and longevity.
  • B-belt trade size with 21/32 in. top width and 13/32 in. thickness
  • 138 in. outside length
  • Wrapped edge construction
  • Resistance to heat and oil
  • Operating temperature range of -22°F to +194°F
  • Reinforced with polyester cords
Catalog No. 50-277-1472 Supplier Grainger Supplier No. 13V799
This B-section classical V-belt features a wrapped construction designed for reliable power transmission in single or multiple V-belt drive applications. Engineered for performance in general industrial settings, the belt is built with polyester reinforcing cords and a rubber compound body to ensure durability and longevity.
  • B-belt trade size with 21/32 in. top width and 13/32 in. thickness
  • 138 in. outside length
  • Wrapped edge construction
  • Resistance to heat and oil
  • Operating temperature range of -22°F to +194°F
  • Reinforced with polyester cords