Learn More
Grainger Dayton Classical V-Belt, Wedge, 164 in Length, 1 ea
Catalog No. 502772706
Encompass_Preferred
This C-section classical wedge V-belt is designed for reliable power transmission in single or multiple V-belt drive systems. Constructed with a durable rubber compound and reinforced with polyester cords, it features a wrapped edge design for enhanced performance and longevity.
  • Industry number C160
  • Outside length of 164 in.
  • Top width of 7/8 in. and 17/32 in. thickness
  • Operating temperature range of -22°F to +194°F
  • Heat-resistant and oil-resistant properties
  • Single-ribbed construction
Catalog No. 50-277-2706 Supplier Grainger Supplier No. 13V839
This C-section classical wedge V-belt is designed for reliable power transmission in single or multiple V-belt drive systems. Constructed with a durable rubber compound and reinforced with polyester cords, it features a wrapped edge design for enhanced performance and longevity.
  • Industry number C160
  • Outside length of 164 in.
  • Top width of 7/8 in. and 17/32 in. thickness
  • Operating temperature range of -22°F to +194°F
  • Heat-resistant and oil-resistant properties
  • Single-ribbed construction