Learn More
Grainger V-Belt Pulley 1/2 Fixed 3.25 OD Iron
Catalog No. 502752819
Encompass_Preferred
The Grainger 14A165 is a Westward 1/4-inch drive socket adapter that converts a 1/4-inch square drive to a 3/8-inch square drive. It is made from chrome vanadium steel for durability and has a black oxide finish to resist corrosion. This socket adapter allows you to use 1/4-inch drive tools with a 3/8-inch drive ratchet, breaker bar or extension bar. It measures 2-1/2 inches long and weighs 0.05 lbs. The Westward 14A165 socket adapter is ideal for mechanics to easily switch between different drive size tools.
Catalog No. 50-275-2819 Supplier Grainger Supplier No. 14A165
The Grainger 14A165 is a Westward 1/4-inch drive socket adapter that converts a 1/4-inch square drive to a 3/8-inch square drive. It is made from chrome vanadium steel for durability and has a black oxide finish to resist corrosion. This socket adapter allows you to use 1/4-inch drive tools with a 3/8-inch drive ratchet, breaker bar or extension bar. It measures 2-1/2 inches long and weighs 0.05 lbs. The Westward 14A165 socket adapter is ideal for mechanics to easily switch between different drive size tools.