Learn More
Grainger V-Belt Pulley 5/8 Fixed 3.25 OD CastIron
Catalog No. 502753524
Encompass_Preferred
The V-Belt Pulley from AK32 is a cast iron pulley with a 2.75" pitch diameter and 0.5" face width. It has a standard A-section v-belt groove and is designed for industrial power transmission applications. The pulley can be mounted on a 0.625" diameter shaft with a set screw. It is rated for up to 1 HP at 1750 RPM. The pulley is made in the USA and conforms to AGMA standards. With its durable cast iron construction, this pulley provides reliable performance for industrial machinery drives.
Catalog No. 50-275-3524 Supplier Grainger Supplier No. 14A166
The V-Belt Pulley from AK32 is a cast iron pulley with a 2.75" pitch diameter and 0.5" face width. It has a standard A-section v-belt groove and is designed for industrial power transmission applications. The pulley can be mounted on a 0.625" diameter shaft with a set screw. It is rated for up to 1 HP at 1750 RPM. The pulley is made in the USA and conforms to AGMA standards. With its durable cast iron construction, this pulley provides reliable performance for industrial machinery drives.