Learn More
Grainger V-Belt Pulley Cast Iron 10.25 O.D.
Catalog No. 502752524
Encompass_Preferred
The TB Wood's V-Belt Pulley is a V-belt sheave with a plain bore and an outside diameter of 4.5". It has a 0.5" bore size and accepts bushings. The pulley is made from cast iron with a black finish. It is designed for industrial power transmission applications and features a crowned face for proper belt alignment. The pulley can be mounted on a shaft with a locking collar or set screws. It is manufactured in the USA by TB Wood's, a leading power transmission component supplier. With its durable cast iron construction, this pulley provides reliable performance for industrial drive systems.
Catalog No. 50-275-2524 Supplier Grainger Supplier No. 145N28
The TB Wood's V-Belt Pulley is a V-belt sheave with a plain bore and an outside diameter of 4.5". It has a 0.5" bore size and accepts bushings. The pulley is made from cast iron with a black finish. It is designed for industrial power transmission applications and features a crowned face for proper belt alignment. The pulley can be mounted on a shaft with a locking collar or set screws. It is manufactured in the USA by TB Wood's, a leading power transmission component supplier. With its durable cast iron construction, this pulley provides reliable performance for industrial drive systems.