Learn More
Grainger V-Belt Pulley Cast Iron 10.25 O.D.
Catalog No. 502748999
Encompass_Preferred
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 2.5 inch outside diameter, a 1.25 inch bore diameter, and a 0.5 inch bushing length. The pulley is made from cast iron with a rough finish and has a set screw for securing the bushing. This pulley is designed for industrial power transmission applications using a V-belt. With its sturdy cast iron construction, bushing bore, and standard V-belt profile, this pulley provides a durable and versatile solution for industrial drive systems.
Catalog No. 50-274-8999 Supplier Grainger Supplier No. 145P84
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 2.5 inch outside diameter, a 1.25 inch bore diameter, and a 0.5 inch bushing length. The pulley is made from cast iron with a rough finish and has a set screw for securing the bushing. This pulley is designed for industrial power transmission applications using a V-belt. With its sturdy cast iron construction, bushing bore, and standard V-belt profile, this pulley provides a durable and versatile solution for industrial drive systems.