Learn More
Grainger V-Belt Pulley Cast Iron 10.75 O.D.
Catalog No. 502749218
Encompass_Preferred
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 3.25 inch outside diameter, a 1.25 inch bore diameter, a 0.5 inch bushing length, and a 0.625 inch bushing inside diameter. The pulley is made from cast iron with a rough finish and operates at speeds up to 3600 RPM. It is designed to be used with fractional horsepower motors in light- to medium-duty applications. This V-belt pulley allows for shaft misalignment and has set screws for securing to the shaft.
Catalog No. 50-274-9218 Supplier Grainger Supplier No. 145P89
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 3.25 inch outside diameter, a 1.25 inch bore diameter, a 0.5 inch bushing length, and a 0.625 inch bushing inside diameter. The pulley is made from cast iron with a rough finish and operates at speeds up to 3600 RPM. It is designed to be used with fractional horsepower motors in light- to medium-duty applications. This V-belt pulley allows for shaft misalignment and has set screws for securing to the shaft.