Learn More
Grainger V-Belt Pulley Cast Iron 10.75 O.D.
Catalog No. 502753476
Encompass_Preferred
The V-Belt Pulley Bushed Bore from Baldor is a V-belt pulley with a bushing bore. It has a 4L section and a 1-1/4 inch bore diameter. The pulley is made of cast iron with a rough finish. It is designed for use with fractional horsepower motors in light industrial applications. The pulley can accommodate a V-belt up to 22/32 inches wide and has a maximum speed rating of 6000 ft/min. It weighs approximately 3 lbs. This V-belt pulley provides a cost-effective power transmission solution.
Catalog No. 50-275-3476 Supplier Grainger Supplier No. 145T42
The V-Belt Pulley Bushed Bore from Baldor is a V-belt pulley with a bushing bore. It has a 4L section and a 1-1/4 inch bore diameter. The pulley is made of cast iron with a rough finish. It is designed for use with fractional horsepower motors in light industrial applications. The pulley can accommodate a V-belt up to 22/32 inches wide and has a maximum speed rating of 6000 ft/min. It weighs approximately 3 lbs. This V-belt pulley provides a cost-effective power transmission solution.