Learn More
Grainger V-Belt Pulley Cast Iron 10.75 O.D.
Catalog No. 502749170
Encompass_Preferred
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 3.25" outside diameter, a 1.25" bore, and a 0.5" bushing. The pulley is made from cast iron with an oil-impregnated bronze bushing. It is designed for industrial power transmission applications using a V-belt. The pulley can accommodate a wide range of bushing bore sizes. It is manufactured in the USA by Browning Manufacturing.
Catalog No. 50-274-9170 Supplier Grainger Supplier No. 145P40
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 3.25" outside diameter, a 1.25" bore, and a 0.5" bushing. The pulley is made from cast iron with an oil-impregnated bronze bushing. It is designed for industrial power transmission applications using a V-belt. The pulley can accommodate a wide range of bushing bore sizes. It is manufactured in the USA by Browning Manufacturing.