Learn More
Grainger V-Belt Pulley Cast Iron 11.25 O.D.
Catalog No. 502749077
Encompass_Preferred
The V-Belt Pulley Bushed Bore 145N30 is a V-belt pulley with a bushing bore. It has a 3.00 inch outside diameter, a 1.25 inch bore diameter, and a 0.50 inch bushing length. The pulley is made of cast iron with an oil-impregnated bronze bushing. It has a QD bushing type and operates at speeds up to 4000 feet per minute. The pulley accepts V-belts sized A30 through A44 and is rated for up to 3 horsepower. This pulley can be used for power transmission applications.
Catalog No. 50-274-9077 Supplier Grainger Supplier No. 145N30
The V-Belt Pulley Bushed Bore 145N30 is a V-belt pulley with a bushing bore. It has a 3.00 inch outside diameter, a 1.25 inch bore diameter, and a 0.50 inch bushing length. The pulley is made of cast iron with an oil-impregnated bronze bushing. It has a QD bushing type and operates at speeds up to 4000 feet per minute. The pulley accepts V-belts sized A30 through A44 and is rated for up to 3 horsepower. This pulley can be used for power transmission applications.