Learn More
Grainger V-Belt Pulley Cast Iron 11.75 O.D.
Catalog No. 502752105
Encompass_Preferred
The V-Belt Pulley Bushed Bore from Baldor Dodge is a V-belt pulley with a bushing bore. It has a 3.25" outside diameter, a 1.25" bore diameter, a 0.5" bushing length, and accepts a 0.5" shaft. The pulley is made from cast iron with an oil-impregnated bronze bushing. It is designed for industrial V-belt power transmission applications. The pulley can be mounted on a shaft using a set screw or bolt. It provides a durable and economical V-belt drive solution.
Catalog No. 50-275-2105 Supplier Grainger Supplier No. 145T55
The V-Belt Pulley Bushed Bore from Baldor Dodge is a V-belt pulley with a bushing bore. It has a 3.25" outside diameter, a 1.25" bore diameter, a 0.5" bushing length, and accepts a 0.5" shaft. The pulley is made from cast iron with an oil-impregnated bronze bushing. It is designed for industrial V-belt power transmission applications. The pulley can be mounted on a shaft using a set screw or bolt. It provides a durable and economical V-belt drive solution.