Learn More
Grainger V-Belt Pulley Cast Iron 14.25 O.D.
Catalog No. 502749223
Encompass_Preferred
The V-Belt Pulley Fixed Bore 145T09 is a fixed bore pulley made by Browning. It has a 3.00" outside diameter, 1.50" bore diameter, 0.50" face width, and 0.19" crown width. The pulley is constructed from cast iron with a rough finish and has a set screw for securing the pulley to a shaft. This pulley is designed for use with V-belts and has a static balance grade of G6.3. It is rated for rotational speeds up to 3600 RPM. The overall dimensions are 3.50" x 3.50" x 1.50". This heavy-duty pulley provides a rigid drive component for industrial power transmission applications.
Catalog No. 50-274-9223 Supplier Grainger Supplier No. 145R04
The V-Belt Pulley Fixed Bore 145T09 is a fixed bore pulley made by Browning. It has a 3.00" outside diameter, 1.50" bore diameter, 0.50" face width, and 0.19" crown width. The pulley is constructed from cast iron with a rough finish and has a set screw for securing the pulley to a shaft. This pulley is designed for use with V-belts and has a static balance grade of G6.3. It is rated for rotational speeds up to 3600 RPM. The overall dimensions are 3.50" x 3.50" x 1.50". This heavy-duty pulley provides a rigid drive component for industrial power transmission applications.