Learn More
Grainger V-Belt Pulley Cast Iron 14.25 O.D.
Catalog No. 502749168
Encompass_Preferred
The Standard V-Belt Pulley Bushed 145N40 is a V-belt pulley with a 4.5" outside diameter and a 1.875" bore. It has a cast iron hub and is made of cast iron with a rough finish. The pulley has a QD bushing installed and is rated for 145 Newton meters of torque. This pulley can be used with v-belts sized A, B, C, and D. It is manufactured by Browning and is made in the USA.
Catalog No. 50-274-9168 Supplier Grainger Supplier No. 145N40
The Standard V-Belt Pulley Bushed 145N40 is a V-belt pulley with a 4.5" outside diameter and a 1.875" bore. It has a cast iron hub and is made of cast iron with a rough finish. The pulley has a QD bushing installed and is rated for 145 Newton meters of torque. This pulley can be used with v-belts sized A, B, C, and D. It is manufactured by Browning and is made in the USA.