Learn More
Grainger V-Belt Pulley Cast Iron 15.25 O.D.
Catalog No. 502749066
Encompass_Preferred
The V-Belt Pulley from Grainger has a bushing bore of 1-1/2 inches, an outside diameter of 5 inches, and a face width of 1-1/4 inches. It is constructed of cast iron with a rough finish and has a standard bushing with setscrew for easy installation. This pulley is designed for industrial power transmission applications using a V-belt. With its sturdy construction, it provides reliable performance for driving motors, fans, pumps and other rotating equipment. The pulley can withstand high radial loads and its large diameter allows efficient power transmission. Overall, this is a heavy-duty V-belt pulley well-suited for industrial settings.
Catalog No. 50-274-9066 Supplier Grainger Supplier No. 145R07
The V-Belt Pulley from Grainger has a bushing bore of 1-1/2 inches, an outside diameter of 5 inches, and a face width of 1-1/4 inches. It is constructed of cast iron with a rough finish and has a standard bushing with setscrew for easy installation. This pulley is designed for industrial power transmission applications using a V-belt. With its sturdy construction, it provides reliable performance for driving motors, fans, pumps and other rotating equipment. The pulley can withstand high radial loads and its large diameter allows efficient power transmission. Overall, this is a heavy-duty V-belt pulley well-suited for industrial settings.