Learn More
Grainger V-Belt Pulley Cast Iron 15.75 O.D.
Catalog No. 502751900
Encompass_Preferred
The V-Belt Pulley from Grainger is a cast iron pulley with an outside diameter of 15.75 inches. It has a 1.125 inch bore and accepts a 1 inch shaft. The pulley has a 0.5 inch face width and operates at speeds up to 6580 feet per minute. It is used with V-belts sized A, B, C, D and E. This heavy duty pulley is ideal for industrial power transmission applications.
Catalog No. 50-275-1900 Supplier Grainger Supplier No. 145M41
The V-Belt Pulley from Grainger is a cast iron pulley with an outside diameter of 15.75 inches. It has a 1.125 inch bore and accepts a 1 inch shaft. The pulley has a 0.5 inch face width and operates at speeds up to 6580 feet per minute. It is used with V-belts sized A, B, C, D and E. This heavy duty pulley is ideal for industrial power transmission applications.