Learn More
Grainger V-Belt Pulley Cast Iron 16.4 O.D.
Catalog No. 502752552
Encompass_Preferred
The V-Belt Pulley Fixed Bore from Grainger is a fixed bore pulley with a V-belt groove. It has a cast iron construction and is black oxide coated. The pulley has a 1-7/16 inch bore size and accepts a 1-7/16 inch shaft. The overall pulley diameter is 4-1/2 inches and the V-belt groove diameter is 3-3/8 inches. This is a fixed bore pulley so it cannot be adjusted. It is well-suited for industrial power transmission applications.
Catalog No. 50-275-2552 Supplier Grainger Supplier No. 145M44
The V-Belt Pulley Fixed Bore from Grainger is a fixed bore pulley with a V-belt groove. It has a cast iron construction and is black oxide coated. The pulley has a 1-7/16 inch bore size and accepts a 1-7/16 inch shaft. The overall pulley diameter is 4-1/2 inches and the V-belt groove diameter is 3-3/8 inches. This is a fixed bore pulley so it cannot be adjusted. It is well-suited for industrial power transmission applications.