Learn More
Grainger V-Belt Pulley Cast Iron 18.25 O.D.
Catalog No. 502749047
Encompass_Preferred
The Standard V-Belt Pulley Fixed from Grainger is a fixed-position pulley designed for use with V-belts. It has a cast iron hub and is made of either cast iron or steel for durability. This pulley has a standard bushing with a 1-inch bore and set screws for securing it to a shaft. It comes in diameters ranging from 3 to 12 inches, with multiple groove options. The pulley can be used for power transmission in industrial machinery when paired with a V-belt. Its all-steel construction provides reliable performance. This heavy-duty pulley is ideal for industrial applications requiring a fixed pulley for a V-belt drive system.
Catalog No. 50-274-9047 Supplier Grainger Supplier No. 145R17
The Standard V-Belt Pulley Fixed from Grainger is a fixed-position pulley designed for use with V-belts. It has a cast iron hub and is made of either cast iron or steel for durability. This pulley has a standard bushing with a 1-inch bore and set screws for securing it to a shaft. It comes in diameters ranging from 3 to 12 inches, with multiple groove options. The pulley can be used for power transmission in industrial machinery when paired with a V-belt. Its all-steel construction provides reliable performance. This heavy-duty pulley is ideal for industrial applications requiring a fixed pulley for a V-belt drive system.