Learn More
Grainger V-Belt Pulley Cast Iron 2.2 O.D.
Catalog No. 502749090
Encompass_Preferred
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 3.25" outside diameter, a 1.25" bore diameter, and a 0.5" bushing length. The pulley is made from cast iron with an oil-impregnated bronze bushing. It is designed for use with fractional horsepower motors in light- to medium-duty applications. This V-belt pulley can be used to change the speed ratio between a driving and driven shaft.
Catalog No. 50-274-9090 Supplier Grainger Supplier No. 145R27
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 3.25" outside diameter, a 1.25" bore diameter, and a 0.5" bushing length. The pulley is made from cast iron with an oil-impregnated bronze bushing. It is designed for use with fractional horsepower motors in light- to medium-duty applications. This V-belt pulley can be used to change the speed ratio between a driving and driven shaft.