Learn More
Grainger V-Belt Pulley Cast Iron 2.6 O.D.
Catalog No. 502752870
Encompass_Preferred
The Grainger V-Belt Pulley is a 3-groove pulley made of cast iron with a bore diameter of 1-1/4 inches. It has a 4-1/2 inch outside diameter and 1-1/8 inch face width. The pulley has a set screw and bushing for easy installation onto a shaft. It is commonly used to drive fans, blowers, conveyors and other industrial equipment using a v-belt drive. This heavy duty pulley provides efficient power transmission.
Catalog No. 50-275-2870 Supplier Grainger Supplier No. 145R38
The Grainger V-Belt Pulley is a 3-groove pulley made of cast iron with a bore diameter of 1-1/4 inches. It has a 4-1/2 inch outside diameter and 1-1/8 inch face width. The pulley has a set screw and bushing for easy installation onto a shaft. It is commonly used to drive fans, blowers, conveyors and other industrial equipment using a v-belt drive. This heavy duty pulley provides efficient power transmission.