Learn More
Grainger V-Belt Pulley Cast Iron 2.8 O.D.
Catalog No. 502752871
Encompass_Preferred
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 3.25 inch outside diameter, a 1.25 inch bore, and a 0.5 inch bushing. The pulley is made from cast iron with an oil-impregnated bronze bushing. It is designed for industrial power transmission applications using a V-belt. The pulley can be mounted on a shaft with a bushing to reduce friction. This is a standard V-belt pulley for basic power transmission tasks.
Catalog No. 50-275-2871 Supplier Grainger Supplier No. 145R45
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 3.25 inch outside diameter, a 1.25 inch bore, and a 0.5 inch bushing. The pulley is made from cast iron with an oil-impregnated bronze bushing. It is designed for industrial power transmission applications using a V-belt. The pulley can be mounted on a shaft with a bushing to reduce friction. This is a standard V-belt pulley for basic power transmission tasks.