Learn More
Grainger V-Belt Pulley Cast Iron 20.35 O.D.
Catalog No. 502752520
Encompass_Preferred
The Grainger V-Belt Pulley is a bushing bore pulley with a 2.25" outside diameter, 1.25" bore diameter, and 0.5" bushing length. It has a cast iron hub construction and is designed for use with v-belts. This pulley can be used for power transmission applications and features set screws for securing it to a shaft. With its compact size, it is well suited for light duty power transmission tasks. The pulley's bushing bore allows for easy installation and replacement. Overall, this is a durable, versatile v-belt pulley for basic power transmission needs.
Catalog No. 50-275-2520 Supplier Grainger Supplier No. 145M55
The Grainger V-Belt Pulley is a bushing bore pulley with a 2.25" outside diameter, 1.25" bore diameter, and 0.5" bushing length. It has a cast iron hub construction and is designed for use with v-belts. This pulley can be used for power transmission applications and features set screws for securing it to a shaft. With its compact size, it is well suited for light duty power transmission tasks. The pulley's bushing bore allows for easy installation and replacement. Overall, this is a durable, versatile v-belt pulley for basic power transmission needs.