Learn More
Grainger V-Belt Pulley Cast Iron 3.05 O.D.
Catalog No. 502749108
Encompass_Preferred
The Grainger 5RHZ9 is a 5/8" fixed v-belt pulley made of cast iron with an outside diameter of 3.05". It has a 0.31" bore and accepts a 5/8" shaft. This pulley can be used for power transmission applications and features a setscrew for securing it to the shaft. With its cast iron construction, it provides durability for industrial use. The fixed v-belt pulley design allows for simple installation and changing of v-belts.
Catalog No. 50-274-9108 Supplier Grainger Supplier No. 145R46
The Grainger 5RHZ9 is a 5/8" fixed v-belt pulley made of cast iron with an outside diameter of 3.05". It has a 0.31" bore and accepts a 5/8" shaft. This pulley can be used for power transmission applications and features a setscrew for securing it to the shaft. With its cast iron construction, it provides durability for industrial use. The fixed v-belt pulley design allows for simple installation and changing of v-belts.