Learn More
Grainger V-Belt Pulley Cast Iron 3.05 O.D.
Catalog No. 502748938
Encompass_Preferred
The V-Belt Pulley Bushed Bore 145N66 is a V-belt pulley with a bushing bore. It has a bore diameter of 1-1/4 inches and an outside diameter of 6 inches. The pulley has a B-section profile and is made of cast iron with an oil-impregnated bronze bushing. It is designed for use with fractional and metric horsepower drives. The pulley can accommodate a wide range of belt sizes and has set screws for securing it to the shaft. This heavy-duty pulley is ideal for industrial power transmission applications.
Catalog No. 50-274-8938 Supplier Grainger Supplier No. 145N66
The V-Belt Pulley Bushed Bore 145N66 is a V-belt pulley with a bushing bore. It has a bore diameter of 1-1/4 inches and an outside diameter of 6 inches. The pulley has a B-section profile and is made of cast iron with an oil-impregnated bronze bushing. It is designed for use with fractional and metric horsepower drives. The pulley can accommodate a wide range of belt sizes and has set screws for securing it to the shaft. This heavy-duty pulley is ideal for industrial power transmission applications.