Learn More
Grainger V-Belt Pulley Cast Iron 3.25 O.D.
Catalog No. 502749109
Encompass_Preferred
The Grainger V-Belt Pulley Fixed Bore 145T09 is a fixed bore pulley for use with v-belts. It has a 3.25 inch outside diameter and a 0.875 inch bore size. The pulley is made from cast iron with a black oxide finish. It is rated for speeds up to 6580 feet per minute. The pulley can be used with v-belts sized A, B, C, and D. It weighs approximately 1.4 pounds. This is a high quality, durable v-belt pulley suitable for industrial applications.
Catalog No. 50-274-9109 Supplier Grainger Supplier No. 145R50
The Grainger V-Belt Pulley Fixed Bore 145T09 is a fixed bore pulley for use with v-belts. It has a 3.25 inch outside diameter and a 0.875 inch bore size. The pulley is made from cast iron with a black oxide finish. It is rated for speeds up to 6580 feet per minute. The pulley can be used with v-belts sized A, B, C, and D. It weighs approximately 1.4 pounds. This is a high quality, durable v-belt pulley suitable for industrial applications.