Learn More
Grainger V-Belt Pulley Cast Iron 3.45 O.D.
Catalog No. 502752877
Encompass_Preferred
The V-Belt Pulley Bushed Bore from Baldor is a V-belt pulley with a bushing bore. It has a 3.25" outside diameter, a 1.25" bore, and a 0.5" bushing. The pulley is made from cast iron with an oil-impregnated bronze bushing. It is designed for use with fractional horsepower motors in light- to medium-duty applications. This V-belt pulley can be used to change the speed ratio between a motor and driven equipment when used with a V-belt. It requires minimal maintenance besides occasional lubrication.
Catalog No. 50-275-2877 Supplier Grainger Supplier No. 145R52
The V-Belt Pulley Bushed Bore from Baldor is a V-belt pulley with a bushing bore. It has a 3.25" outside diameter, a 1.25" bore, and a 0.5" bushing. The pulley is made from cast iron with an oil-impregnated bronze bushing. It is designed for use with fractional horsepower motors in light- to medium-duty applications. This V-belt pulley can be used to change the speed ratio between a motor and driven equipment when used with a V-belt. It requires minimal maintenance besides occasional lubrication.