Learn More
Grainger V-Belt Pulley Cast Iron 3.55 O.D.
Catalog No. 502753477
Encompass_Preferred
The V-Belt Pulley Fixed Bore from Grainger is a fixed bore pulley for use with V-belts. It has a cast iron construction with a painted finish. The pulley has a fixed 1-7/16 inch bore and is available in multiple belt sizes from A to E with diameters ranging from 2 to 12 inches. The maximum RPM rating is 3600. This heavy duty pulley can be used for power transmission applications.
Catalog No. 50-275-3477 Supplier Grainger Supplier No. 145U07
The V-Belt Pulley Fixed Bore from Grainger is a fixed bore pulley for use with V-belts. It has a cast iron construction with a painted finish. The pulley has a fixed 1-7/16 inch bore and is available in multiple belt sizes from A to E with diameters ranging from 2 to 12 inches. The maximum RPM rating is 3600. This heavy duty pulley can be used for power transmission applications.