Learn More
Grainger V-Belt Pulley Cast Iron 3.55 O.D.
Catalog No. 502749113
Encompass_Preferred
The Grainger 5-8" Fixed V-Belt Pulley is a cast iron pulley with a 5" outside diameter and an 8" face width. It has a fixed bore size of 3-55/64", is rated for a maximum speed of 3600 RPM, and is designed for use with v-belts. This heavy duty pulley provides high load capacity and vibration dampening. It can be used for power transmission in industrial machinery.
Catalog No. 50-274-9113 Supplier Grainger Supplier No. 145R58
The Grainger 5-8" Fixed V-Belt Pulley is a cast iron pulley with a 5" outside diameter and an 8" face width. It has a fixed bore size of 3-55/64", is rated for a maximum speed of 3600 RPM, and is designed for use with v-belts. This heavy duty pulley provides high load capacity and vibration dampening. It can be used for power transmission in industrial machinery.