Learn More
Grainger V-Belt Pulley Cast Iron 3.75 O.D.
Catalog No. 502749169
Encompass_Preferred
The V-Belt Pulley Cast Iron 3.75" O.D. from Grainger is a cast iron pulley with an outside diameter of 3.75 inches. It has a 0.75 inch bore and accepts a 0.75 inch shaft. The pulley has a V-belt groove and is used for power transmission applications. This pulley is made from durable cast iron and can be used with v-belts up to a 38 inch outer circumference. The maximum RPM rating is 1725 RPM. Overall, this is a sturdy cast iron v-belt pulley for industrial power transmission uses.
Catalog No. 50-274-9169 Supplier Grainger Supplier No. 145N81
The V-Belt Pulley Cast Iron 3.75" O.D. from Grainger is a cast iron pulley with an outside diameter of 3.75 inches. It has a 0.75 inch bore and accepts a 0.75 inch shaft. The pulley has a V-belt groove and is used for power transmission applications. This pulley is made from durable cast iron and can be used with v-belts up to a 38 inch outer circumference. The maximum RPM rating is 1725 RPM. Overall, this is a sturdy cast iron v-belt pulley for industrial power transmission uses.