Learn More
Grainger V-Belt Pulley Cast Iron 3.95 O.D.
Catalog No. 502748942
Encompass_Preferred
The V-Belt Pulley BK62-145U49 is a V-belt pulley made by Browning. It has a 4.5 inch outside diameter and 1.45 inch bore diameter. The pulley has a B section V-belt groove and is made of cast iron with a black oxide finish. This pulley can be used for power transmission applications and with v-belts that have a top width of 0.62 inches. The pulley has set screws for securing it to a shaft.
Catalog No. 50-274-8942 Supplier Grainger Supplier No. 145N85
The V-Belt Pulley BK62-145U49 is a V-belt pulley made by Browning. It has a 4.5 inch outside diameter and 1.45 inch bore diameter. The pulley has a B section V-belt groove and is made of cast iron with a black oxide finish. This pulley can be used for power transmission applications and with v-belts that have a top width of 0.62 inches. The pulley has set screws for securing it to a shaft.