Learn More
Grainger V-Belt Pulley Cast Iron 4.45 O.D.
Catalog No. 502752078
Encompass_Preferred
The Grainger Approved V-Belt Pulley is a high-quality pulley designed for use with V-belts. It has a cast iron construction for durability and is precision-machined for accurate grooves and bore. The pulley has a standard cross-section for use with common V-belts and is available in diameters from 2" to 12" with bores ranging from 5/8" to 2-3/8". It can be used for power transmission in industrial machinery. This heavy-duty pulley provides efficient power transfer for industrial applications.
Catalog No. 50-275-2078 Supplier Grainger Supplier No. 145U17
The Grainger Approved V-Belt Pulley is a high-quality pulley designed for use with V-belts. It has a cast iron construction for durability and is precision-machined for accurate grooves and bore. The pulley has a standard cross-section for use with common V-belts and is available in diameters from 2" to 12" with bores ranging from 5/8" to 2-3/8". It can be used for power transmission in industrial machinery. This heavy-duty pulley provides efficient power transfer for industrial applications.