Learn More
Grainger V-Belt Pulley Cast Iron 4.45 O.D.
Catalog No. 502752106
Encompass_Preferred
The 2AK56 V-Belt Pulley from Grainger is a V-belt sheave made of cast iron with a black finish. It has a 2.25 inch outside diameter, 0.875 inch bore diameter, and 0.5 inch face width. The pulley has a standard b-section v-belt groove and operates at speeds up to 6580 feet per minute. It is designed for use with fractional horsepower motors in light-duty power transmission applications. The pulley is manufactured in the USA and sold individually.
Catalog No. 50-275-2106 Supplier Grainger Supplier No. 145U12
The 2AK56 V-Belt Pulley from Grainger is a V-belt sheave made of cast iron with a black finish. It has a 2.25 inch outside diameter, 0.875 inch bore diameter, and 0.5 inch face width. The pulley has a standard b-section v-belt groove and operates at speeds up to 6580 feet per minute. It is designed for use with fractional horsepower motors in light-duty power transmission applications. The pulley is manufactured in the USA and sold individually.