Learn More
Grainger V-Belt Pulley Cast Iron 4.75 O.D.
Catalog No. 502749078
Encompass_Preferred
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 3.5 inch outside diameter, a 1.25 inch bore, and a 0.5 inch bushing. The pulley is made from cast iron with a black oxide finish. It has a QD bushing and is designed for use with fractional horsepower motors. The pulley can accommodate V-belts sized A, B, C, and D. It is rated for speeds up to 3600 RPM. This pulley provides an economical way to drive and transfer power from motors to pumps, fans, and other equipment.
Catalog No. 50-274-9078 Supplier Grainger Supplier No. 145P01
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 3.5 inch outside diameter, a 1.25 inch bore, and a 0.5 inch bushing. The pulley is made from cast iron with a black oxide finish. It has a QD bushing and is designed for use with fractional horsepower motors. The pulley can accommodate V-belts sized A, B, C, and D. It is rated for speeds up to 3600 RPM. This pulley provides an economical way to drive and transfer power from motors to pumps, fans, and other equipment.