Learn More
Grainger V-Belt Pulley Cast Iron 4.95 O.D.
Catalog No. 502748939
Encompass_Preferred
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 3L V-belt section and a 1-1/16" bore diameter. The pulley is made of cast iron with a black oxide finish. It is designed for industrial power transmission applications and features a setscrew for securing to the shaft. This is a high quality, durable V-belt pulley well-suited for industrial use.
Catalog No. 50-274-8939 Supplier Grainger Supplier No. 145P04
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 3L V-belt section and a 1-1/16" bore diameter. The pulley is made of cast iron with a black oxide finish. It is designed for industrial power transmission applications and features a setscrew for securing to the shaft. This is a high quality, durable V-belt pulley well-suited for industrial use.