Learn More
Grainger V-Belt Pulley Cast Iron 5.25 O.D.
Catalog No. 502748940
Encompass_Preferred
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 3.5 inch outside diameter, a 1.5 inch bore diameter, and a 0.5 inch bushing length. The pulley is made from cast iron with a rough finish and has a 0.25 inch face width. It is designed for use with fractional horsepower drives. The pulley can accommodate a V-belt with a cross section of A, B, C, D, or E. It provides a means to transfer rotational force from motors to driven equipment via a V-belt.
Catalog No. 50-274-8940 Supplier Grainger Supplier No. 145P08
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 3.5 inch outside diameter, a 1.5 inch bore diameter, and a 0.5 inch bushing length. The pulley is made from cast iron with a rough finish and has a 0.25 inch face width. It is designed for use with fractional horsepower drives. The pulley can accommodate a V-belt with a cross section of A, B, C, D, or E. It provides a means to transfer rotational force from motors to driven equipment via a V-belt.