Learn More
Grainger V-Belt Pulley Cast Iron 5.25 O.D.
Catalog No. 502750554
Encompass_Preferred
The V-Belt Pulley Fixed Bore 145R78 is a fixed bore pulley made by Browning. It has a 3.25 inch outside diameter, 1.25 inch bore diameter, and 0.5 inch face width. The pulley is made from cast iron with a rough finish and is designed for use with v-belts. It has set screws for securing to a shaft. This pulley can be used for power transmission applications.
Catalog No. 50-275-0554 Supplier Grainger Supplier No. 145R78
The V-Belt Pulley Fixed Bore 145R78 is a fixed bore pulley made by Browning. It has a 3.25 inch outside diameter, 1.25 inch bore diameter, and 0.5 inch face width. The pulley is made from cast iron with a rough finish and is designed for use with v-belts. It has set screws for securing to a shaft. This pulley can be used for power transmission applications.