Learn More
Grainger V-Belt Pulley Cast Iron 5.25 O.D.
Catalog No. 502750555
Encompass_Preferred
The V-Belt Pulley Bushed Bore 145R79 is a V-belt pulley with a bushing bore. It has a 1-3/8" bore diameter and is made of cast iron. The pulley has a 4L cross section and is 7" in outside diameter. It is designed for use with fractional horsepower drives. The pulley can accommodate a V-belt with a top width up to 1/2". It is manufactured in the USA by Browning.
Catalog No. 50-275-0555 Supplier Grainger Supplier No. 145R79
The V-Belt Pulley Bushed Bore 145R79 is a V-belt pulley with a bushing bore. It has a 1-3/8" bore diameter and is made of cast iron. The pulley has a 4L cross section and is 7" in outside diameter. It is designed for use with fractional horsepower drives. The pulley can accommodate a V-belt with a top width up to 1/2". It is manufactured in the USA by Browning.