Learn More
Grainger V-Belt Pulley Cast Iron 5.25 O.D.
Catalog No. 502748950
Encompass_Preferred
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 3.25 inch outside diameter, a 1.25 inch bore, and a 0.5 inch bushing. The pulley is made from cast iron with an oil-impregnated bronze bushing. It is used with fractional horsepower motors in light- to medium-duty applications. This V-belt pulley allows for shaft misalignment and has a maximum speed of 3600 RPM. It works with A, B, and C section v-belts.
Catalog No. 50-274-8950 Supplier Grainger Supplier No. 145U27
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 3.25 inch outside diameter, a 1.25 inch bore, and a 0.5 inch bushing. The pulley is made from cast iron with an oil-impregnated bronze bushing. It is used with fractional horsepower motors in light- to medium-duty applications. This V-belt pulley allows for shaft misalignment and has a maximum speed of 3600 RPM. It works with A, B, and C section v-belts.