Learn More
Grainger V-Belt Pulley Cast Iron 5.25 O.D.
Catalog No. 502749080
Encompass_Preferred
The V-Belt Pulley BK85 from Grainger is a cast iron pulley with a 5" outside diameter and a 1-7/16" bore. It has a V-belt groove and is used to transmit power between the driving and the driven shaft. This pulley can be mounted on shafts with a keyway and secured with a setscrew. It is rated for industrial V-belt drives under 1 hp and 1725 rpm. The overall width of the pulley is 1-3/4". This is a standard v-belt pulley for light-duty power transmission applications.
Catalog No. 50-274-9080 Supplier Grainger Supplier No. 145U26
The V-Belt Pulley BK85 from Grainger is a cast iron pulley with a 5" outside diameter and a 1-7/16" bore. It has a V-belt groove and is used to transmit power between the driving and the driven shaft. This pulley can be mounted on shafts with a keyway and secured with a setscrew. It is rated for industrial V-belt drives under 1 hp and 1725 rpm. The overall width of the pulley is 1-3/4". This is a standard v-belt pulley for light-duty power transmission applications.