Learn More
Grainger V-Belt Pulley Cast Iron 5.25 O.D.
Catalog No. 502749075
Encompass_Preferred
The Dayton V-Belt Pulley is a 3.5 inch diameter cast iron pulley with a 1 inch bore and a V-belt groove. It has set screws for securing to a shaft. The pulley can be used to drive or be driven by a V-belt in power transmission applications. It is rated for up to 3 horsepower at 1750 RPM. The pulley is made in the USA by Dayton, a leading manufacturer of electric motors, power transmission, material handling and maintenance products. This is a high quality pulley well suited for industrial applications requiring power transmission via V-belts.
Catalog No. 50-274-9075 Supplier Grainger Supplier No. 145U22
The Dayton V-Belt Pulley is a 3.5 inch diameter cast iron pulley with a 1 inch bore and a V-belt groove. It has set screws for securing to a shaft. The pulley can be used to drive or be driven by a V-belt in power transmission applications. It is rated for up to 3 horsepower at 1750 RPM. The pulley is made in the USA by Dayton, a leading manufacturer of electric motors, power transmission, material handling and maintenance products. This is a high quality pulley well suited for industrial applications requiring power transmission via V-belts.