Learn More
Grainger V-Belt Pulley Cast Iron 5.45 O.D.
Catalog No. 502752020
Encompass_Preferred
The V-Belt Pulley BK62-145U49 is a V-belt pulley made by Browning. It has a 4.5 inch outside diameter and 1.45 inch bore diameter. The pulley has a B section V-belt groove and is made of cast iron with a black oxide finish. This pulley can be used for power transmission applications and with v-belts that fit the B section profile. The pulley has set screws for securing it to a shaft. It is rated for up to 1750 RPM and 1 to 15 HP. This heavy duty pulley provides reliable performance for industrial power transmission uses.
Catalog No. 50-275-2020 Supplier Grainger Supplier No. 145U29
The V-Belt Pulley BK62-145U49 is a V-belt pulley made by Browning. It has a 4.5 inch outside diameter and 1.45 inch bore diameter. The pulley has a B section V-belt groove and is made of cast iron with a black oxide finish. This pulley can be used for power transmission applications and with v-belts that fit the B section profile. The pulley has set screws for securing it to a shaft. It is rated for up to 1750 RPM and 1 to 15 HP. This heavy duty pulley provides reliable performance for industrial power transmission uses.