Learn More
Grainger V-Belt Pulley Cast Iron 5.45 O.D.
Catalog No. 502753475
Encompass_Preferred
The Grainger website features a wide selection of V-belt pulleys in various sizes, materials, and configurations. These pulleys are designed to transmit rotary motion between shafts using V-belts. They feature machined grooves to match common V-belt sizes. Pulley options include cast iron, steel, nylon, and other materials. Diameters range from 1" to over 12". Some feature multiple grooves, adjustable pitch diameter, taper bushings, and other features. Overall this page provides a comprehensive overview of industrial V-belt pulleys that could be used for power transmission in machinery and equipment.
Catalog No. 50-275-3475 Supplier Grainger Supplier No. 145U30
The Grainger website features a wide selection of V-belt pulleys in various sizes, materials, and configurations. These pulleys are designed to transmit rotary motion between shafts using V-belts. They feature machined grooves to match common V-belt sizes. Pulley options include cast iron, steel, nylon, and other materials. Diameters range from 1" to over 12". Some feature multiple grooves, adjustable pitch diameter, taper bushings, and other features. Overall this page provides a comprehensive overview of industrial V-belt pulleys that could be used for power transmission in machinery and equipment.