Learn More
Grainger V-Belt Pulley Cast Iron 5.45 O.D.
Catalog No. 502749056
Encompass_Preferred
The V-Belt Pulley 2AK56 from Grainger is a cast iron pulley with a 2 inch bore and a 5 inch outside diameter. It has a standard V-belt groove and is used with fractional and metric horsepower drives. This pulley can be mounted on shafts, jackshafts, countershafts and more to transfer rotational force. It is commonly used in industrial power transmission applications. The pulley is made in the USA and conforms to ANSI standards. It weighs approximately 2 pounds.
Catalog No. 50-274-9056 Supplier Grainger Supplier No. 145P12
The V-Belt Pulley 2AK56 from Grainger is a cast iron pulley with a 2 inch bore and a 5 inch outside diameter. It has a standard V-belt groove and is used with fractional and metric horsepower drives. This pulley can be mounted on shafts, jackshafts, countershafts and more to transfer rotational force. It is commonly used in industrial power transmission applications. The pulley is made in the USA and conforms to ANSI standards. It weighs approximately 2 pounds.